Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 3-Way Stopcocks
Product Code. 10088170
Change view
Click to view available options
Plug Bore:
2.5 mm
4 mm
Includes:
PTFE Plug, Screw Thread Retaining Device
Solid Glass Plug, Screw Thread Retaining Device
Solid Glass Plug, Screw Thread Rubber Retaining Device
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore Includes unitSize
10088170 4 mm PTFE Plug, Screw Thread Retaining Device Each
10777851 2.5 mm Solid Glass Plug, Screw Thread Retaining Device Each
10509971 2.5 mm PTFE Plug, Screw Thread Retaining Device Each
10612472 4 mm Solid Glass Plug, Screw Thread Retaining Device Each
10597911 4 mm Solid Glass Plug, Screw Thread Rubber Retaining Device Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10088170 Supplier Lenz Laborglasinstrumente™ Supplier No. 02542404
Only null left
Add to basket
Add to basket
Specifications
Type 3-way Stopcock
Plug Bore 4 mm
Material Borosilicate Glass
Certifications/Compliance DIN 12554
Includes PTFE Plug, Screw Thread Retaining Device