missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)aniline, 97%, Thermo Scientific™

Product Code. 10089161
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10089161 10 g Amber Glass Bottle 10g
10344712 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10089161 Supplier Thermo Scientific Maybridge Supplier No. CC21914EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89353-42-4
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD03407309
InChI Key PJGDCPOPSNUYHC-UHFFFAOYSA-N
Synonym 3-1h-pyrrol-1-yl aniline, 3-pyrrol-1-yl-phenylamine, 3-pyrrol-1-yl aniline, 3-pyrrolylphenylamine, 3-1-pyrrolyl aniline, 3-pyrrol-1-yl-aniline, 1-3-aminophenyl pyrrole, benzenamine, 3-1h-pyrrol-1-yl
PubChem CID 2760546
IUPAC Name 3-pyrrol-1-ylaniline
SMILES C1=CN(C=C1)C2=CC(=CC=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 3-(1H-pyrrol-1-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.