missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%

Product Code. 10021044 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021044 1 g Glass bottle 1g
10366364 5 g Glass bottle 5g
11985541 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10021044 Supplier Thermo Scientific Acros Supplier No. 395020010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98737-29-2 | C15H21NO3 | 263.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98737-29-2
Molecular Formula C15H21NO3
Molecular Weight (g/mol) 263.34
MDL Number MFCD02258997
InChI Key NVPOUMXZERMIJK-QWHCGFSZSA-N
Synonym 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane, 2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane, tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate, 2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate, tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate, tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate, 2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane, ksc523o6b
PubChem CID 9903372
IUPAC Name tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 125.0°C to 127.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: <0.1% (20°C)
Formula Weight 263.34
Percent Purity 98%
Chemical Name or Material (2S, 3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
compliance-icons
Product Identifier
  • (2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.