missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-1-Chloro-3-(1H-indol-4-yloxy)propan-2-ol, Mikromol™

Product Code. 30049673
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049673 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049673 Supplier Mikromol Supplier No. MM0082.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-1-Chloro-3-(1H-indol-4-yloxy)propan-2-ol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-1-Chloro-3-(1H-indol-4-yloxy)propan-2-ol
CAS 130115-66-1
Analyte or Component Names (2RS)-1-Chloro-3-(1H-indol-4-yloxy)propan-2-ol
Shipping Condition Room Temperature
Molecular Formula C11 H12 Cl N O2
Certifications/Compliance ISO 9001
Synonym (2RS)-1-Chloro-3-(1H-indol-4-yloxy)propan-2-ol, Pindolol Imp. F (EP), Pindolol Impurity F
InChI Formula InChI=1S/C11H12ClNO2/c12-6-8(14)7-15-11-3-1-2-10-9(11)4-5-13-10/h1-5,8,13-14H,6-7H2
SMILES OC(CCl)COc1cccc2[nH]ccc12
IUPAC Name 1-chloro-3-(1H-indol-4-yloxy)propan-2-ol
Molecular Weight (g/mol) 225.67
Quantity 50 mg
Formula Weight 225.0557 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.