missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-1-[4-[[(2-Isopropoxyethoxy)methoxy]methyl]phenoxy]-3-isopropylaminopropan-2-ol, Mikromol™

Product Code. 30049502
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30049502 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30049502

Brand: Mikromol MM0460.21

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-1-[4-[[(2-Isopropoxyethoxy)methoxy]methyl]phenoxy]-3-isopropylaminopropan-2-ol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-1-[4-[[(2-Isopropoxyethoxy)methoxy]methyl]phenoxy]-3-isopropylaminopropan-2-ol
CAS 1215342-36-1
Analyte or Component Names (2RS)-1-[4-[[(2-Isopropoxyethoxy)methoxy]methyl]phenoxy]-3-isopropylaminopropan-2-ol
Shipping Condition Room Temperature
Molecular Formula C19 H33 N O5
Certifications/Compliance ISO 9001
Synonym Bisoprolol Fumarate Imp. G (EP), Bisoprolol Imp. G (EP), (2RS)-1-[4-[[(2-Isopropoxyethoxy)methoxy]methyl]phenoxy]-3-isopropylaminopropan-2-ol, 2-Propanol, 1-[4-[[[2-(1-methylethoxy)ethoxy]methoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-, Bisoprolol Related Compound D, Bisoprolol Fumarate Impurity G, Bisoprolol Impurity G
InChI Formula InChI=1S/C19H33NO5/c1-15(2)20-11-18(21)13-25-19-7-5-17(6-8-19)12-23-14-22-9-10-24-16(3)4/h5-8,15-16,18,20-21H,9-14H2,1-4H3
SMILES CC(C)NCC(O)COc1ccc(COCOCCOC(C)C)cc1
IUPAC Name 1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethoxymethyl)phenoxy]propan-2-ol
Molecular Weight (g/mol) 355.47
Quantity 100 mg
Formula Weight 355.2359 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.