missing translation for 'onlineSavingsMsg'
Learn More

HUBER Insulated Stainless Steel Bath 215B

Product Code. 10048614
Change view
Click to view available options
Model:
215 B
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
10048614 215 B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10048614 Supplier HUBER Supplier No. 6012
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to operate to a maximum temperature of +200° C

Insulated stainless steel bath 215B, 15L

Specifications

Capacity (Metric) 15 L
Temperature Range 200°C Max.
Model 215 B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.