missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ISO Certificate, Pore Size: 10 mm and Larger

Product Code. 10136701 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
10 mm
11.20 mm
12.50 mm
13.20 mm
14 mm
16 mm
18 mm
19 mm
20 mm
22.40 mm
25 mm
26.50 mm
28 mm
31.50 mm
35.50 mm
37.50 mm
40 mm
45 mm
50 mm
53 mm
56 mm
63 mm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10136701 31.50 mm Each
10340812 106 mm Each
10085071 11.20 mm Each
10729892 112 mm Each
10729702 100 mm Each
10487001 10 mm Each
10156411 12.50 mm Each
10487571 125 mm Each
10209171 13.20 mm Each
10729322 18 mm Each
10024971 19 mm Each
10320292 14 mm Each
10506032 22.40 mm Each
10690452 25 mm Each
10064931 16 mm Each
10600112 20 mm Each
10126701 26.50 mm Each
10085001 28 mm Each
10055051 37.50 mm Each
10760473 35.50 mm Each
10146931 40 mm Each
10065051 45 mm Each
10147121 50 mm Each
10466431 53 mm Each
10466621 56 mm Each
10044911 63 mm Each
10761623 71 mm Each
10137171 75 mm Each
10610642 90 mm Each
10004941 80 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
This item is not returnable. View return policy
Product Code. 10136701 Supplier RETSCH Supplier No. 60.149.031500
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 1Each
Pore Size 31.50 mm
Type Test Sieve
Certifications/Compliance ISO 3310/1
Material Stainless Steel
For Use With (Equipment) Shaker AS 300
Height (Metric) 50.8mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 203.2 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.