Learn More
BOLA 2-Way Stopcocks
Description
- 2-way stopcock with straight bore and two connections or 3-way stopcock with T-shaped bore and three connections
- With nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
- Conical stopcock plug, tightness is increased by turning the nut on the lower side.
- 3-way stopcock plug with T-shaped mark of flow direction.
- Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Fits Tubing ID | 8 thread |
| Closure Size | M18 |
| Certifications/Compliance | FDA |