missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propylheptanoic acid, 97%

Product Code. 15453888
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453888 250 mg 250mg
15443888 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453888 Supplier Thermo Scientific Alfa Aesar Supplier No. H57414.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31080-39-4 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31080-39-4
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD01723758
InChI Key RXGPYPPCEXISOV-UHFFFAOYNA-N
Synonym 2-propylheptanic acid, 4-nonanecarboxylic acid, heptanoic acid,2-propyl, heptanoic acid, 2-propyl, 2-propylheptansaeure german, 2-n-propylheptanoic acid, 2-n-propyl-1-heptanoic acid, 2-propylheptansaeure, 2-02-00-00313 beilstein handbook reference
PubChem CID 35728
IUPAC Name 2-propylheptanoic acid
SMILES CCCCCC(CCC)C(O)=O

Specifications

Color Colorless to Yellow
Quantity 250 mg
Formula Weight 172.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-n-Propylheptanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.