missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15234308 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15234308 5 g 5g
15224308 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15234308 Supplier Thermo Scientific Acros Supplier No. 453580050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1257535-05-9
Molecular Formula C7H5ClF3NO2S
Molecular Weight (g/mol) 259.63
MDL Number MFCD14583150
InChI Key BNCNLCKWRCXBQH-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine
PubChem CID 51342031
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
SMILES CC1=C(C=CC(=N1)C(F)(F)F)S(Cl)(=O)=O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 259.64
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
compliance-icons
Product Identifier
  • 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.