missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanol, 99+%, Thermo Scientific Chemicals

Product Code. p-86663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367600 100 mL Glass bottle 100mL
10307560 1 L Glass bottle 1 L
10003773 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10367600 Supplier Thermo Scientific Acros Supplier No. 180961000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-07-3
Molecular Weight (g/mol) 80.51
MDL Number MFCD00002829
InChI Key SZIFAVKTNFCBPC-UHFFFAOYSA-N
Synonym ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
PubChem CID 34
ChEBI CHEBI:28200

Specifications

Acidity 0.2mg KOH/g max.
CAS Min % 99.0
CAS Max % 100.0
Melting Point -63.0°C
Color Colorless
Density 1.2000g/mL
Boiling Point 129.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C2H5ClO
Linear Formula ClCH2CH2OH
Refractive Index 1.4405 to 1.4425
Quantity 100 mL
Beilstein 01, 337
Fieser 05,117
Merck Index 15, 3847
Specific Gravity 1.2
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,soluble in most organic liquids,insoluble in alifatic hydrocarbons
SMILES C(CCl)O
IUPAC Name 2-chloroethanol
Viscosity 3.4 mPa.s (20°C)
Formula Weight 80.51
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Chloroethanol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H330-Fatal if inhaled.
  • H310-Fatal in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.