missing translation for 'onlineSavingsMsg'
Learn More

2-[(Chloroacetyl)amino]benzoic Acid, TRC

Product Code. 30346872
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30346872

Brand: Toronto Research Chemicals TRCB41833825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(Chloroacetyl)amino]benzoic Acid
CAS 14422-49-2
Molecular Formula C9H8NO3Cl
Quantity 25 mg
Synonym 2-[(2-Chloroacetyl)amino]benzoic Acid, 2-(2-Chloroacetamido)benzoic Acid, 2-(Chloroacetamido)benzoic Acid, 2-Chloroacetylaminobenzoic Acid, N-(Chloroacetyl)anthranilic Acid, N-(chloroacetyl)antranilic Acid
InChI Formula InChI=1S/C9H8ClNO3/c10-5-8(12)11-7-4-2-1-3-6(7)9(13)14/h1-4H,5H2,(H,11,12)(H,13,14)
SMILES OC(=O)c1ccccc1NC(=O)CCl
IUPAC Name 2-[(2-chloroacetyl)amino]benzoic acid
Molecular Weight (g/mol) 213.61
Formula Weight 213.0193
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.