missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[(Chloroacetyl)amino]benzoic Acid, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 2-[(Chloroacetyl)amino]benzoic Acid |
| CAS | 14422-49-2 |
| Molecular Formula | C9H8NO3Cl |
| Quantity | 50 mg |
| Synonym | 2-[(2-Chloroacetyl)amino]benzoic Acid, 2-(2-Chloroacetamido)benzoic Acid, 2-(Chloroacetamido)benzoic Acid, 2-Chloroacetylaminobenzoic Acid, N-(Chloroacetyl)anthranilic Acid, N-(chloroacetyl)antranilic Acid |
| InChI Formula | InChI=1S/C9H8ClNO3/c10-5-8(12)11-7-4-2-1-3-6(7)9(13)14/h1-4H,5H2,(H,11,12)(H,13,14) |
| SMILES | OC(=O)c1ccccc1NC(=O)CCl |
| IUPAC Name | 2-[(2-chloroacetyl)amino]benzoic acid |
| Molecular Weight (g/mol) | 213.61 |
| Formula Weight | 213.0193 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction