missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-beta-nitrostyrene, 98%

Product Code. 11442677
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442677 5 g 5g
11452677 25 g 25g
11462677 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442677 Supplier Thermo Scientific Alfa Aesar Supplier No. A14882.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3156-34-1 | C8H6ClNO2 | 183.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3156-34-1
Molecular Formula C8H6ClNO2
Molecular Weight (g/mol) 183.591
MDL Number MFCD00024820
InChI Key QHKJTRDWAZGBLR-AATRIKPKSA-N
Synonym 1-chloro-2-2-nitrovinyl benzene, 2-chloro-omega-nitrostyrene, 2-chloro-b-nitrostyrene, e-1-chloro-2-2-nitrovinyl benzene, 1-chloro-2-e-2-nitrovinyl benzene, trans-2-chloro-beta-nitrostyrene, 2-chloro-beta-nitrostyrene, styrene, o-chloro-.beta.-nitro, benzene,1-chloro-2-2-nitroethenyl, 1-chloro-2-e-2-nitroethenyl benzene
PubChem CID 5369313
IUPAC Name 1-chloro-2-[(E)-2-nitroethenyl]benzene
SMILES C1=CC=C(C(=C1)C=C[N+](=O)[O-])Cl

Specifications

Melting Point 46°C to 49°C
Quantity 5 g
Beilstein 2328095
Formula Weight 183.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-beta-nitrostyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.