missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid, Thermo Scientific™

Product Code. 10048219
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10048219 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10048219 Supplier Thermo Scientific Maybridge Supplier No. MAY00114DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 590371-81-6
Packaging Amber Glass Bottle
Molecular Formula C7H3ClF3NO2
MDL Number MFCD08458125
Quantity 1 g
Synonym 2-chloro-4-trifluoromethyl nicotinic acid, 2-chloro-4-trifluoromethyl pyridine-3-carboxylic acid, 3-carboxy-2-chloro-4-trifluoromethyl pyridine, acmc-1az7h, ksc269c2b, 3-pyridinecarboxylicacid, 2-chloro-4-trifluoromethyl, 3-pyridinecarboxylic acid, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethyl-3-pyridinecarboxylic acid
InChI Key FZIJHEOXLKCIRZ-UHFFFAOYSA-N
SMILES OC(=O)C1=C(C=CN=C1Cl)C(F)(F)F
IUPAC Name 2-chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol) 225.55
PubChem CID 11770138
Formula Weight 225.55
Percent Purity 97%
Chemical Name or Material 2-chloro-4-(trifluoromethyl)pyridine-3-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.