missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,5-dimethylpyrazine, 97%, Thermo Scientific™

Product Code. 10153753
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10153753 5 g 5g
10397663 1 g 1g
10613235 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10153753 Supplier Thermo Scientific Maybridge Supplier No. MO08011DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 38557-72-1
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00126945
InChI Key BTGGHNHGPURMEO-UHFFFAOYSA-N
PubChem CID 11672680
IUPAC Name 2-chloro-3,5-dimethylpyrazine
SMILES CC1=CN=C(Cl)C(C)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Synonym 2-chloro 3,5-dimethyl pyarazine, 2-chloro-3,5-dimethyl-1,4-diazine, 2-chloro-3,5-dimethyl-pyrazine, 3,5-dimethyl-2-chloropyrazine, 2-chloro 3,5-dimethyl pyrazine, 2-chloro-3,5-dimethyl pyrazine, pyrazine,2-chloro-3,5-dimethyl, 2-chloranyl-3,5-dimethyl-pyrazine, pyrazine, 2-chloro-3,5-dimethyl, 3-chloro-2,6-dimethylpyrazine
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-3, 5-dimethylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.