missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Carboxybenzoyl Amlodipine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-([2-([(4RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy)ethyl]carbamoyl)benzoic Acid |
| CAS | 318465-73-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 H29 Cl N2 O8 |
| Quantity | 50 mg |
| Synonym | 2-[[2-[[(4RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]carbamoyl]benzoic Acid, Amlodipine Besilate Imp. H (EP) |
| InChI Formula | InChI=1S/C28H29ClN2O8/c1-4-39-28(36)24-21(15-38-14-13-30-25(32)17-9-5-6-10-18(17)26(33)34)31-16(2)22(27(35)37-3)23(24)19-11-7-8-12-20(19)29/h5-12,23,31H,4,13-15H2,1-3H3,(H,30,32)(H,33,34) |
| SMILES | CCOC(=O)C1=C(COCCNC(=O)c2ccccc2C(=O)O)NC(=C(C1c3ccccc3Cl)C(=O)OC)C |
| IUPAC Name | 2-[2-[[4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethylcarbamoyl]benzoic acid |
| Molecular Weight (g/mol) | 556.99 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?