missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-pyridinemethanol, 95%

Product Code. 15487898
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487898 1 g 1g
15497898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487898 Supplier Thermo Scientific Alfa Aesar Supplier No. H61375.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122306-01-8 | C6H6BrNO | 188.024 g/mol

2-Bromo-5-pyridinemethanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-pyridinemethanol is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122306-01-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD04974508
InChI Key QPPDKOIDAYZUHN-UHFFFAOYSA-N
Synonym 6-bromopyridin-3-yl methanol, 2-bromo-5-hydroxymethyl pyridine, 6-bromo-pyridin-3-yl-methanol, 6-bromo-3-pyridinemethanol, 6-bromo-pyridin-3-yl methanol, 2-bromo-pyridin-5-yl-methanol, 6-bromo-3-pyridyl methanol, 2-bromo-5-hydroxymethylpyridine, 6-bromo-3-pyridinyl methanol, 6-bromopyridine-3-methanol
PubChem CID 6421249
IUPAC Name (6-bromopyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)Br

Specifications

Melting Point 48°C to 51°C
Quantity 1 g
Formula Weight 188.02
Percent Purity 95%
Chemical Name or Material 2-Bromo-5-pyridinemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.