missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoyloxy-4,6-dihydroxybenzaldehyde, TRC

Product Code. 30328760
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328760 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB20833510MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 2-Benzoyloxy-4,6-dihydroxybenzaldehyde
CAS 32292-78-7
Molecular Formula C14H10O5
Quantity 10 mg
Synonym 2,4,6-Trihydroxybenzaldehyde 2-Benzoate, 2-(Benzoyloxy)-4,6-dihydroxybenzaldehyde
InChI Formula InChI=1S/C14H10O5/c15-8-11-12(17)6-10(16)7-13(11)19-14(18)9-4-2-1-3-5-9/h1-8,16-17H
SMILES Oc1cc(O)c(C=O)c(OC(=O)c2ccccc2)c1
IUPAC Name (2-formyl-3,5-dihydroxyphenyl) benzoate
Molecular Weight (g/mol) 258.23
Formula Weight 258.0528
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.