missing translation for 'onlineSavingsMsg'
Learn More

2'-Amino-2'-deoxyuridine, 98%, Thermo Scientific™

Product Code. p-7043163
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
15497129 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 15497129

Brand: Thermo Scientific Alfa Aesar J65923.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Amino-2′-deoxyuridine is used in the synthesis of anticode oligonucleotide, ribozyme and in antisense. It also acts as an intermediate and used in chemical research.

Solubility
Partly soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26889-39-4
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.219
MDL Number MFCD01317284
InChI Key LLIPTMWIZVIUSX-XVFCMESISA-N
Synonym 2'-amino-2'-deoxyuridine, 2'-amino-2'-deoxy-uridine, uridine, 2'-amino-2'-deoxy, 1-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, 2'-amino,2'-deoxyuridine, 2'-amino-2'-deoxy uridine, 2'-deoxy-2'-amino-uridine, 2'-amino-2'-deoxy-d-uridine, 2,4 1h,3h-pyrimidinedione, 1-2-amino-2-deoxy-b-d-ribofuranosyl
PubChem CID 6482479
IUPAC Name 1-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)N

Specifications

Color White to Yellow
Quantity 1 g
Solubility Information Partly soluble in water.
Formula Weight 243.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Amino-2'-deoxyuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.