missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-methylbenzimidazole, 98+%

Product Code. 10147713 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147713 10 g Glass bottle 10g
10318910 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10147713 Supplier Thermo Scientific Acros Supplier No. undefined
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1622-57-7 | C8H9N3 | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1622-57-7
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD00142855
InChI Key XDFZKQJLNGNJAN-UHFFFAOYSA-N
Synonym 2-amino-1-methylbenzimidazole, 1-methyl-1h-benzimidazol-2-amine, 1-methyl-1h-benzo d imidazol-2-amine, 1-methyl-2-aminobenzimidazole, 1h-benzimidazol-2-amine, 1-methyl, 1-methyl-1h-1,3-benzodiazol-2-amine, ccris 4355, 2-amino-1-methyl-benzimidazole, 1h-benzimidazol-2-amine,1-methyl, 1-methyl-1h-benzoimidazol-2-ylamine
PubChem CID 74187
IUPAC Name 1-methylbenzimidazol-2-amine
SMILES CN1C(N)=NC2=CC=CC=C12

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 203°C to 206°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 147.18
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Amino-1-methylbenzimidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.