missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethoxy]benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10102844
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10102844 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10102844 Supplier Thermo Scientific Maybridge Supplier No. CC51316DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 206261-63-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H14N2O
MDL Number MFCD08690291
Quantity 1 g
Synonym 2-2-dimethylamino ethoxy benzonitrile, 2-2-dimethylaminoethyloxy benzonitrile, 2-2-dimethylamino-ethoxy-benzonitrile, benzonitrile,2-2-dimethylamino ethoxy, 2-2-dimethylamino ethoxy benzenecarbonitrile
InChI Key DFZYVQTZWFEMTN-UHFFFAOYSA-N
SMILES CN(C)CCOC1=CC=CC=C1C#N
IUPAC Name 2-[2-(dimethylamino)ethoxy]benzonitrile
Molecular Weight (g/mol) 190.246
PubChem CID 10330161
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 2-[2-(Dimethylamino)ethoxy]benzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.