missing translation for 'onlineSavingsMsg'
Learn More

[[[[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetyl]oxy]acetic Acid (Diacetic Aceclofenac), Mikromol™

Product Code. 18031480
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18031480 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18031480 Supplier Mikromol Supplier No. MM0181.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[[[[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetyl]oxy]acetic Acid (Diacetic Aceclofenac)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diacetic Aceclofenac
CAS 1216495-92-9
Analyte or Component Names Diacetic Aceclofenac
Shipping Condition Room Temperature
Molecular Formula C20 H17 Cl2 N O8
Certifications/Compliance ISO 9001
Synonym Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, carboxymethyl ester (9CI), 2-[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetoxy]acetic acid, Aceclofar, Airtal, Biofenac, Bristaflam, Falcol, Gerbin, PR 82/3, Tresquim, [[[[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetyl]oxy]acetic Acid (Diacetic Aceclofenac), [[[[[[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetyl]oxy]acetyl]oxy]acetic Acid, Aceclofenac Imp. H (EP), Diacetic Aceclofenac, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid 2-[2-(carboxymethoxy)-2-oxoethoxy]-2-oxoethyl ester, Aceclofenac Impurity H
InChI Formula InChI=1S/C20H17Cl2NO8/c21-13-5-3-6-14(22)20(13)23-15-7-2-1-4-12(15)8-17(26)30-10-19(28)31-11-18(27)29-9-16(24)25/h1-7,23H,8-11H2,(H,24,25)
SMILES OC(=O)COC(=O)COC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name 2-[2-[2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetyl]oxyacetyl]oxyacetic acid
Molecular Weight (g/mol) 470.26
Quantity 50 mg
Formula Weight 469.0331 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.