missing translation for 'onlineSavingsMsg'
Learn More

2-[[2-[[(4RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]carbamoyl]benzoic Acid, LoGiCal

Product Code. 30460743
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460743 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460743 Supplier LoGiCal Supplier No. LGCFOR0383.11

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

2-[[2-[[(4RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]carbamoyl]benzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-([2-([(4 RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy)ethyl]carbamoyl)benzoic Acid
CAS 318465-73-5
Analyte or Component Names 2-([2-([(4 RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy)ethyl]carbamoyl)benzoic Acid
Shipping Condition Room Temperature
Molecular Formula C28 H29 Cl N2 O8
Certifications/Compliance ISO 9001
Synonym Amlodipine Besilate Imp. H (EP), Amlodipine Imp. H (EP), 2-[[2-[[(4 RS)-4-(2-Chlorophenyl)-3-(ethoxycarbonyl)-5-(methoxycarbonyl)-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethyl]carbamoyl]benzoic Acid, Amlodipine Besilate Impurity H, Amlodipine Impurity H
InChI Formula InChI=1 S/C28H29ClN2O8/c1-4-39-28(36)24-21(15-38-14-13-30-25(32)17-9-5-6-10-18(17)26(33)34)31-16(2)22(27(35)37-3)23(24)19-11-7-8-12-20(19)29/h5-12,23,31 H,4,13-15H2,1-3H3,(H,30,32)(H,33,34)
SMILES CCOC(=O)C1=C(COCCNC(=O)c2ccccc2C(=O)O)NC(=C(C1c3ccccc3Cl)C(=O)OC)C
IUPAC Name 2-[2-[[4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethylcarbamoyl]benzoic acid
Industry Type Forensics
Molecular Weight (g/mol) 556.99
Quantity 10 mg
Formula Weight 556.1612 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.