missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-1,2-Cyclohexanedimethanol, TRC

Product Code. 30365450
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30365450 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365450 Supplier Toronto Research Chemicals Supplier No. TRCC987000100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S,2S)-1,2-Cyclohexanedimethanol
CAS 3205-34-3
Molecular Formula C8H16O2
Quantity 100 g
Synonym (+)-trans-1,2-Cyclohexanedimethanol, (1S-trans)-1,2-Cyclohexanedimethanol, (1S,2S)-1,2-Bis(hydroxymethyl)cyclohexane, (1S,2S)-Cyclohexane-1,2-dimethanol, (S,S)-1,2-Bis(hydroxymethyl)cyclohexane, trans-(+)-1,2-Bis(hydroxymethyl)cyclohexane
InChI Formula InChI=1S/C8H16O2/c9-5-7-3-1-2-4-8(7)6-10/h7-10H,1-6H2/t7-,8-/m1/s1
SMILES OC[C@@H](CCCC1)[C@H]1CO
IUPAC Name [(1S,2S)-2-(hydroxymethyl)cyclohexyl]methanol
Molecular Weight (g/mol) 144.21
Formula Weight 144.115
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.