missing translation for 'onlineSavingsMsg'
Learn More

(1RS,2SR)-2-[(1,3-Dioxoisoindolin-2-yl)methyl]-N,N-diethyl-1-phenylcyclopropane-1-carboxamide, Mikromol

Product Code. 30459658
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30459658 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459658 Supplier Mikromol Supplier No. MM0746.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1RS,2SR)-2-[(1,3-Dioxoisoindolin-2-yl)methyl]-N,N-diethyl-1-phenylcyclopropane-1-carboxamide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1RS,2SR)-2-((1,3-Dioxoisoindolin-2-yl)methyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide
CAS 105310-75-6
Analyte or Component Names (1RS,2SR)-2-((1,3-Dioxoisoindolin-2-yl)methyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide
Shipping Condition Room Temperature
Molecular Formula C23 H24 N2 O3
Certifications/Compliance ISO 9001
Synonym Cyclopropanecarboxamide, 2-[(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-N,N-diethyl-1-phenyl-, (1R,2S)-rel-, Cyclopropanecarboxamide, 2-[(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-N,N-diethyl-1-phenyl-, cis-, rel-(1R,2S)-2-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]-N,N-diethyl-1-phenylcyclopropanecarboxamide, (1RS,2SR)-2-[(1,3-Dioxoisoindolin-2-yl)methyl]-N,N-diethyl-1-phenylcyclopropane-1-carboxamide
InChI Formula InChI=1S/C23H24N2O3/c1-3-24(4-2)22(28)23(16-10-6-5-7-11-16)14-17(23)15-25-20(26)18-12-8-9-13-19(18)21(25)27/h5-13,17H,3-4,14-15H2,1-2H3/t17-,23+/m0/s1
Solution Type Neat
SMILES CCN(CC)C(=O)[C@]1(C[C@H]1CN2C(=O)c3ccccc3C2=O)c4ccccc4
IUPAC Name (1S,2R)-2-[(1,3-dioxoisoindol-2-yl)methyl]-N,N-diethyl-1-phenylcyclopropane-1-carboxamide
Molecular Weight (g/mol) 376.45
Quantity 25 mg
Formula Weight 376.1787 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.