missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 1R,3R-RSL3
GREENER_CHOICE

Product Code. 18775526
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18775526 10 mg 10mg
18738466 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18775526 Supplier Tocris Bioscience™ Supplier No. 6687/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Negative control for 1S,3R-RSL3 (Cat. No. 6118)

1R,3R-RSL3 is a negative control for 1S,3R-RSL3 (Cat. No. 6118).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1 R,3 R)-Methyl 2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1-[4-(methoxycarbonyl)phenyl]-1 H-pyrido[3,4-b]indole-3-carboxylate
CAS 1219810-15-7
Quantity 10 mg
Target Glutathione Peroxidase 4/GPX4
Molecular Formula C23H21ClN2O5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.