missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%

Product Code. 10195410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10195410 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10195410 Supplier Thermo Scientific Acros Supplier No. 294662500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118628-68-5 | C16H20N2 | 240.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 118628-68-5
CAS Min % 98.5
CAS Max % 100.0
Melting Point 48.0°C to 51.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C16H20N2
MDL Number MFCD00216955
Quantity 250 mg
Fieser 16,159; 16,159; 16,153; 16,153; 17,127
Specific Rotation Condition + 49.50 (20.00°C c=2,MeOH)
Specific Rotation + 49.50
Synonym 1r,2r-n1,n2-dimethyl-1,2-diphenylethane-1,2-diamine, 1r,2r-n,n'-dimethyl-1,2-diphenyl-1,2-ethanediamine, methyl 1r,2r-2-methylamino-1,2-diphenylethyl amine, 1r,2r-n,n'-dimethyl-1,2-diphenyl-1,2-ethylenediamine, 1r,2r-n,n'-dimethyl-1,2-diphenylethylenediamine, 1r,2r-n,n/'-dimethyl-1,2-diphenylethylenediamine, 1r,2r-n,n'-dimethyl-1,2-diphenylethane-1,2-diamine, 1r,2r-n,n'-dimethyl-1,2-diphenyl-ethane-1,2-diamine, 1r,2r-n∼1∼,n∼2∼-dimethyl-1,2-diphenylethane-1,2-diamine
InChI Key BTHYVQUNQHWNLS-HZPDHXFCSA-N
SMILES CNC(C1=CC=CC=C1)C(C2=CC=CC=C2)NC
IUPAC Name (1R,2R)-N,N'-dimethyl-1,2-diphenylethane-1,2-diamine
Molecular Weight (g/mol) 240.35
PubChem CID 2724997
Formula Weight 240.35
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material (1R,2R)-(+)-N,N′-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.