missing translation for 'onlineSavingsMsg'
Learn More

18β-Glycyrrhetinic acid, MedChemExpress

Product Code. p-200101080
Encompass_Preferred
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
This item is not returnable. View return policy

Product Code. 16451328

Brand: Medchem Express HYN0180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 18β-Glycyrrhetinic acid is the major bioactive component of Glycyrrhizae Radix and possesses anti-ulcerative, anti-inflammatory and antiproliferative properties.

18β-Glycyrrhetinic acid is the major bioactive component of Glycyrrhizae Radix and possesses anti-ulcerative, anti-inflammatory and antiproliferative properties.

Specifications

Chemical Name or Material 18β-Glycyrrhetinic acid
CAS 471-53-4
Color White
Purity Grade Notes Research
Molecular Formula C30H46O4
Quantity 5 g
Solubility Information DMSO : ≥ 250 mg/mL (531.15 mM)
SMILES CC1(C)[C@@H](O)CC[C@]([C@@]1([H])CC[C@@]([C@@]2(CC[C@]3(CC[C@](C(O)=O)(C[C@]3(C2=C4)[H])C)C)C)5C)(C)[C@@]5([H])C4=O
Molecular Weight (g/mol) 470.68
Formula Weight 470.68
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.