missing translation for 'onlineSavingsMsg'
Learn More

17Alpha-Methyl-5Beta-estrane-3Alpha,17Beta-diol, TRC

Product Code. 30474375
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474375 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474375 Supplier Toronto Research Chemicals Supplier No. TRCM40581510MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17Alpha-Methyl-5Beta-estrane-3Alpha,17Beta-diol
CAS 31658-49-8
Molecular Formula C19 H32 O2
Quantity 10 mg
Synonym 17α-Methyl-5β-estrane-3α,17β-diol, (3R,5R,8R,9R,10S,13S,14S,17S)-13,17-Dimethylhexadecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol, 17ALPHA-METHYL-5BETA-ESTRANE-3ALPHA,17BETA-DIOL
InChI Formula InChI=1S/C19H32O2/c1-18-9-7-15-14-6-4-13(20)11-12(14)3-5-16(15)17(18)8-10-19(18,2)21/h12-17,20-21H,3-11H2,1-2H3/t12-,13-,14+,15-,16-,17+,18+,19+/m1/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@@H]4[C@H]3CC[C@]12C
IUPAC Name (3R,5R,8R,9R,10S,13S,14S,17S)-13,17-dimethyl-2,3,4,5,6,7,8,9,10,11,12,14,15,16-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 292.46
Formula Weight 292.2402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.