Learn More
Lenz Laborglasintrumente 1-way Stopcock with PTFE Key Bore
Product Code. 10132141
Change view
Click to view available options
Plug Bore:
8 mm
Unit Size:
Each
Product Code. Plug Bore unitSize
10132141 8 mm Each
1 options
Product Code. 10132141 Supplier Lenz Laborglasintrumente Supplier No. 02162408
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Stopcocks, straight bore, with PTFE plug
  • Acc. to DIN 12541-1
  • With PTFE plug and screwthread retaining device
  • One straight bore
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Plug Bore 8 mm
Material Borosilicate Glass (3.3)