missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10170922
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10170922 10 g Amber Glass Bottle 10g
10222642 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10170922 Supplier Thermo Scientific Maybridge Supplier No. GK01501EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 116344-17-3
Molecular Formula C13H14N2O2
Molecular Weight (g/mol) 230.267
MDL Number MFCD00068140
InChI Key CBYITWPSSDTYTN-UHFFFAOYSA-N
PubChem CID 2777578
IUPAC Name 1-phenyl-5-propylpyrazole-4-carboxylic acid
SMILES CCCC1=C(C=NN1C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 1-phenyl-5-propyl-1h-pyrazole-4-carboxylic acid, 1h-pyrazole-4-carboxylicacid, 1-phenyl-5-propyl, maybridge1_003832, pubchem14158, acmc-209xta, 1-phenyl-5-propyl-1h-pyrazole-4-carboxylicacid, buttpark 36\18-32, 1h-pyrazole-4-carboxylicacid,1-phenyl-5-propyl, 1h-pyrazole-4-carboxylic acid, 1-phenyl-5-propyl
Formula Weight 230.27
Percent Purity 97%
Chemical Name or Material 1-Phenyl-5-propyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.