missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

Product Code. 10116033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10116033 100 g Glass bottle 100g
10024523 25 g Glass bottle 25g
11914571 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10116033 Supplier Thermo Scientific Acros Supplier No. 396651000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22767-49-3 | C5H11NaO3S | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007541,MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
PubChem CID 23664617
IUPAC Name sodium;pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Absorbance 0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. C=0.5 M in water at 220nm, 0.1 max. (C=0.5 M in water) at 210nm
Melting Point >300.0°C
Color White
Loss on Drying 2% (150°C) max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)4SO3Na
Quantity 100 g
Solubility Information Solubility in water: soluble.
Formula Weight 174.2
Percent Purity 99+%
Grade Ion Pair Chromatography
Physical Form Crystalline Powder
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • 1-Pentanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.