missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl butyrate, 98%

Product Code. 10746622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10746622 1 g Glass bottle 1g
10787002 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10746622 Supplier Thermo Scientific Acros Supplier No. 334950010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3121-70-8 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3121-70-8
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00003923
InChI Key XIVJNRXPRQKFRZ-UHFFFAOYSA-N
Synonym 1-naphthyl butyrate, .alpha.-naphthyl butyrate, alpha-naphthyl butyrate, naphthalen-1-yl butyrate, butanoic acid, 1-naphthalenyl ester, butyric acid, 1-naphthyl ester, 1-naphthylbutyrate, unii-9uzl9u0cik, butyric acid 1-naphthyl ester, 9uzl9u0cik
PubChem CID 76571
IUPAC Name naphthalen-1-yl butanoate
SMILES CCCC(=O)OC1=CC=CC2=CC=CC=C21

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.0900g/mL
Boiling Point 125°C to 130°C (1.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible in water
Formula Weight 214.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Naphthyl butyrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.