missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Ferrocenylethanol, 99%, Thermo Scientific Chemicals

Product Code. 11436860
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436860 1 g 1g
11446860 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11436860 Supplier Thermo Scientific Alfa Aesar Supplier No. L19681.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

(±)-1-Ferrocenylethanol is a ligand in titanium-catalyzed intermolecular hydroamination of terminal alkynes (Markovnikov vs. anti-Markovnikov addition). Used as a reactant for single-electron transfer living radical polymerization reactions, preparation of ferrocene-modified thiopyrimidines as anticancer agents, preparation of rersible addition-fragmentation chain transfer (RAFT) agent for RAFT polymerization of styrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-1-Ferrocenylethanol is a ligand in titanium-catalyzed intermolecular hydroamination of terminal alkynes (Markovnikov vs. anti-Markovnikov addition). Used as a reactant for single-electron transfer living radical polymerization reactions, preparation of ferrocene-modified thiopyrimidines as anticancer agents, preparation of rersible addition-fragmentation chain transfer (RAFT) agent for RAFT polymerization of styrene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1277-49-2
Molecular Formula C12H14FeO
Molecular Weight (g/mol) 230.09
MDL Number MFCD00013752
InChI Key KGIBJAFJTDOHBV-LURJTMIESA-N
Synonym 1-Hydroxyethylferrocene; alpha-Methylferrocenemethanol
IUPAC Name (±)-1-Ferrocenylethanol
SMILES [Fe].c1cccc1.CC(O)c1cccc1

Specifications

Melting Point 77°C to 79°C
Flash Point >100°C (212°F)
Quantity 1 g
Formula Weight 230.09
Percent Purity 99%
Chemical Name or Material (+/-)-1-Ferrocenylethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.