missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctane 99%, ACROS Organics™

Product Code. 10041750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041750 100 mL Glass bottle 100mL
10428482 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041750 Supplier Thermo Scientific Acros Supplier No. 154711000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 111-85-3
Molecular Formula C8H17Cl
Molecular Weight (g/mol) 148.674
MDL Number MFCD00001022
InChI Key CNDHHGUSRIZDSL-UHFFFAOYSA-N
Synonym n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543
PubChem CID 8142
IUPAC Name 1-chlorooctane
SMILES CCCCCCCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)7Cl
Refractive Index 1.4288 to 1.4308 (20°C, 589nm)
Quantity 100 mL
Beilstein 01, 159
Formula Weight 148.68
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1-Chlorooctane, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.