missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine, Mikromol™

Product Code. 17902046
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
17902046 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 17902046

Brand: Mikromol MM0677.16

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
CAS 120013-45-8
Analyte or Component Names 1-Benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
Shipping Condition Room Temperature
Molecular Formula C24 H29 N O2
Certifications/Compliance ISO 9001
Synonym 1-Benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine, 4-[(5,6-Dimethoxy-1H-inden-2-yl)methyl]-1-(phenylmethyl)piperidine, 1-Benzyl-4-(5,6-dimethoxy-1H-2-indenylmethyl)piperidine, Donepezil Impurity
InChI Formula InChI=1S/C24H29NO2/c1-26-23-15-21-13-20(14-22(21)16-24(23)27-2)12-18-8-10-25(11-9-18)17-19-6-4-3-5-7-19/h3-7,13,15-16,18H,8-12,14,17H2,1-2H3
SMILES COc1cc2CC(=Cc2cc1OC)CC3CCN(Cc4ccccc4)CC3
IUPAC Name 1-benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
Molecular Weight (g/mol) 363.49
Quantity 50 mg
Formula Weight 363.2198 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.