missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-2-methylindoline Hydrochloride, Mikromol™

Product Code. 17931726
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931726 100 mg 100mg
17951026 25 mg 25mg
30462131 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17931726 Supplier Mikromol Supplier No. MM0477.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Amino-2-methylindoline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Amino-2-methylindoline Hydrochloride
CAS 31529-47-2
Analyte or Component Names 1-Amino-2-methylindoline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H12 N2 . Cl H
Certifications/Compliance ISO 17025
Synonym Indapamide Imp. C (EP), 1H-Indol-1-amine, 2,3-dihydro-2-methyl-, hydrochloride (1:1), 1H-Indol-1-amine, 2,3-dihydro-2-methyl-, monohydrochloride (9CI), Indoline, 1-amino-2-methyl-, monohydrochloride (8CI), 1-Amino-2-methylindoline Monohydrochloride, 2,3-Dihydro-2-methyl-1H-indol-1-amine monohydrochloride, Indapamide Related Compound B as Hydrochloride, Indapamide Impurity C as Hydrochloride
InChI Formula InChI=1S/C9H12N2.ClH/c1-7-6-8-4-2-3-5-9(8)11(7)10;/h2-5,7H,6,10H2,1H3;1H
SMILES Cl.CC1Cc2ccccc2N1N
IUPAC Name 2-methyl-2,3-dihydroindol-1-amine;hydrochloride
Molecular Weight (g/mol) 184.67
Quantity 100 mg
Formula Weight 184.0767 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.