missing translation for 'onlineSavingsMsg'
Learn More

[(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]methanesulfonic Acid Sodium Salt (4-N-Desmethylmetamizole Sodium Salt), Mikromol™

Product Code. 17993046
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17993046

Brand: Mikromol MM0052.110025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]methanesulfonic Acid Sodium Salt (4-N-Desmethylmetamizole Sodium Salt)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ((1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino)methanesulfonic Acid Sodium Salt
CAS 129-89-5
Analyte or Component Names ((1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino)methanesulfonic Acid Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C12 H14 N3 O4 S . Na
Certifications/Compliance ISO 9001
Synonym [(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)amino]methanesulfonic Acid Sodium Salt, 4-N-Desmethylmetamizole Sodium Salt, Melaminsulfone Sodium Salt, Metamizol Sodium Imp. E (EP) as Sodium Salt, Metamizole Sodium Monohydrate Impurity E as Sodium Salt, Metamizole Impurity E as Sodium Salt
InChI Formula InChI=1S/C12H15N3O4S.Na/c1-9-11(13-8-20(17,18)19)12(16)15(14(9)2)10-6-4-3-5-7-10;/h3-7,13H,8H2,1-2H3,(H,17,18,19);/q;+1/p-1
SMILES [Na+].CN1N(C(=O)C(=C1C)NCS(=O)(=O)[O-])c2ccccc2
IUPAC Name sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)amino]methanesulfonate
Molecular Weight (g/mol) 319.31
Quantity 25 mg
Formula Weight 319.0603 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.