missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propylene-d6 Thiourea, TRC

Product Code. 30137230
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137230 1 mg 1mg
30132594 2 mg 2mg
17967509 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30137230 Supplier Toronto Research Chemicals Supplier No. TRCT3113721MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219802-05-7
Molecular Formula C4 D6 H2 N2 S
Molecular Weight (g/mol) 122.2217
Synonym 1,3-Propylene-d6 Thiourea, Tetrahydropyrimidine-2(1H)-thione-4,4,5,5,6,6-D6, Cyclic propylene-d6 thiourea, Trimethylenethiourea-d6, 2-Mercapto-1,4,5,6-tetrahydropyrimidine-4,4,5,5,6,6-d6
IUPAC Name 4,4,5,5,6,6-hexadeuterio-1,3-diazinane-2-thione
SMILES [2H]C1([2H])NC(=S)NC([2H])([2H])C1([2H])[2H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C4H8N2S/c7-4-5-2-1-3-6-4/h1-3H2,(H2,5,6,7)/i1D2,2D2,3D2
Formula Weight 122.0785
Chemical Name or Material 1,3-Propylene-d6 Thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.