missing translation for 'onlineSavingsMsg'
Learn More

1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate, 98%, Thermo Scientific™

Product Code. 11354384
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354384 10 g 10g
11364384 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354384 Supplier Thermo Scientific Alfa Aesar Supplier No. L00956.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54770-27-3
Molecular Formula C38H32Br2P2
Molecular Weight (g/mol) 710.43
MDL Number MFCD00040548
InChI Key TVFYBHGFLHYJNV-GLDAUVFXSA-L
Synonym triphenyl e-2-triphenylphosphonio ethenyl phosphonium dibromide
PubChem CID 123134366
SMILES [Br-].[Br-].C(=C/[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)\[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼236°C to 250°C
Quantity 10 g
Beilstein 3583793
Sensitivity Hygroscopic
IUPAC Name triphenyl-[(E)-2-triphenylphosphaniumylethenyl]phosphanium;dihydrobromide
Formula Weight 728.45 (710.44 Anhydrous)
Percent Purity 98%
Chemical Name or Material 1,2-Vinylenebis(triphenylphosphonium bromide) monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.