missing translation for 'onlineSavingsMsg'
Learn More

1,12-Diaminododecane, 98+%

Product Code. p-7044050
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316866 5 g 5g
11326866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11316866 Supplier Thermo Scientific Alfa Aesar Supplier No. L04258.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2783-17-7 | C12H28N2 | 200.37 g/mol

1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Diaminododecane is used for synthesis of medicinal drugs, polymers.

Solubility
Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2783-17-7
Molecular Formula C12H28N2
Molecular Weight (g/mol) 200.37
MDL Number MFCD00008155
InChI Key QFTYSVGGYOXFRQ-UHFFFAOYSA-N
Synonym 1,12-diaminododecane, 1,12-dodecanediamine, dodecamethylenediamine, dodecyldiamine, 1,12-dodecamethylenediamine, 1,12-diamindodecane, 1,12'-dodecylenediamine, 1,12'-dodecamethylenediamine, unii-j3lm80w9nt, 1,12-dodecylenediamine
PubChem CID 17727
ChEBI CHEBI:49385
IUPAC Name dodecane-1,12-diamine
SMILES C(CCCCCCN)CCCCCN

Specifications

Melting Point 68°C to 72°C
Flash Point 155°C (311°F)
Linear Formula H2N(CH2)12NH2
Quantity 5 g
UN Number UN3259
Beilstein 1742765
Solubility Information Soluble in ethanol.
Formula Weight 200.37
Percent Purity ≥98%
Chemical Name or Material 1,12-Diaminododecane
compliance-icons compliance-icons
Product Identifier
  • 1,12-Diaminododecane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,12-Dodecanediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.