missing translation for 'onlineSavingsMsg'
Learn More

1-(1,3-Benzodioxol-5-yl)-2-bromoethan-1-one, Thermo Scientific™

Product Code. 10133214
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133214 10 g Amber Glass Bottle 10g
10722103 250 mg Amber Glass Bottle 250mg
10511692 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10133214 Supplier Thermo Scientific Maybridge Supplier No. CC01605EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40288-65-1
Molecular Formula C9H7BrO3
Molecular Weight (g/mol) 243.056
InChI Key QBXCVQVFPVXAGS-UHFFFAOYSA-N
PubChem CID 243777
IUPAC Name 1-(1,3-benzodioxol-5-yl)-2-bromoethanone
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 1-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 1-1,3-benzodioxol-5-yl-2-bromoethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethan-1-one, 5-bromoacetyl-1,3-benzodioxolane, ethanone,1-1,3-benzodioxol-5-yl-2-bromo, 1-benzo d 1,3 dioxol-5-yl-2-bromoethanone, 1-benzo 1,3 dioxol-5-yl-2-bromo-ethanone, 1-2h-1,3-benzodioxol-5-yl-2-bromoethanone, ethanone, 1-1,3-benzodioxol-5-yl-2-bromo, 1-2h-benzo 3,4-d 1,3-dioxolen-5-yl-2-bromoethan-1-one
Formula Weight 243.06
Percent Purity 97%
Chemical Name or Material 1-(1, 3-Benzodioxol-5-yl)-2-bromoethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.