Use promo code "25339" to get your promotional price.
Silver Nitrate, Certified AR for Analysis, Fisher Chemical™
Use promo code "25339" to get your promotional price.
| Quantity | 100 g |
|---|
Chemical Identifiers
| CAS | 7761-88-8 |
|---|---|
| Molecular Formula | AgNO3 |
| Molecular Weight (g/mol) | 169.87 |
| MDL Number | MFCD00003414 |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| SMILES | [Ag+].[O-][N+]([O-])=O |
Specification
| Boiling Point | 444°C |
|---|---|
| Melting Point | 212°C |
| CAS | 7761-88-8 |
| Color | White |
| pH | 5.4 to 6.4 |
| Physical Form | Solid |
| Quantity | 100 g |
| Molecular Formula | AgNO3 |
| MDL Number | MFCD00003414 |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| Synonym | silver nitrate,silvernitrate,lunar caustic,silbernitrat,argenti nitras,nitrate d'argent,nitric acid silver i salt,silver mononitrate,silver i nitrate,argerol |
| InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
| SMILES | [Ag+].[O-][N+]([O-])=O |
| IUPAC Name | silver;nitrate |
| Molecular Weight (g/mol) | 169.87 |
| PubChem CID | 24470 |
| ChEBI | CHEBI:32130 |
| Formula Weight | 169.87g/mol |
| Packaging | Glass Jar |
Technical documents
[Silver nitrate]
[Danger]
[{code=OX. SOL. 2, sequence=1, description=Oxidising solid Category 2}, {code=MET. CORR. 1, sequence=2, description=Substances and mixtures corrosive to metals Category 1}, {code=SKIN CORR. 1B, sequence=3, description=Skin corrosion/irritation Category 1 B}, {code=EYE DAM. 1, sequence=4, description=Serious eye damage/eye irritation Category 1}, {code=AQUATIC ACUTE 1, sequence=5, description=ACUTE AQUATIC Acute 1}, {code=AQUA CHRONIC 1, sequence=6, description=LONG-TERM AQUATIC HAZARD Chronic 1}]
[{code=H272, sequence=1, description=May intensify fire; oxidiser.}, {code=H290, sequence=2, description=May be corrosive to metals.}, {code=H314, sequence=3, description=Causes severe skin burns and eye damage.}, {code=H410, sequence=4, description=Very toxic to aquatic life with long lasting effects.}]
[{code=P220, sequence=51, description=Keep/Store away from clothing/combustible materials.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P301+P330+P331, sequence=53, description=IF SWALLOWED: rinse mouth. Do NOT induce vomiting.}, {code=P303+P361+P353, sequence=54, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=55, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P310, sequence=56, description=Immediately call a POISON CENTER/doctor/.}]
[{code=GHS03, sequence=1, description=FLAME OVER CIRCLE}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS09, sequence=3, description=ENVIRONMENT}]
Product Identifier
- Silver nitrate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H272- May intensify fire; oxidiser.
- H290- May be corrosive to metals.
- H314- Causes severe skin burns and eye damage.
- H410- Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P220- Keep/Store away from clothing/combustible materials.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331- IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.