Cyanoacetic acid, 97%
CAS: 372-09-8 | C3H3NO2 | 85.062 g/mol
Overview
319.99 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 372-09-8 Weight: 5.00000kg Transport: UN number : 3261 Chem class : 8 Pack group : II
Quantity 5000 g
Disclaimers: RUO – Research Use Only
Product Code. 11478883
Quantity Price
1 £ 320.0 / 5000g
In stock, ready to ship: 4
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 372-09-8
Molecular Formula C3H3NO2
Molecular Weight (g/mol) 85.062
MDL Number MFCD00002677
InChI Key MLIREBYILWEBDM-UHFFFAOYSA-N
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
PubChem CID 9740
ChEBI CHEBI:51889
IUPAC Name 2-cyanoacetic acid
SMILES C(C#N)C(=O)O
Specification
Chemical Name or Material Cyanoacetic acid
CAS 372-09-8
Melting Point 64°C to 69°C
pH 1.5
Boiling Point 108°C (15 mmHg)
Flash Point 107°C (224°F)
Odor Odorless
Molecular Formula C3H3NO2
MDL Number MFCD00002677
Quantity 5000 g
UN Number UN3261
Beilstein 506325
Sensitivity Hygroscopic
Merck Index 14,2689
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Synonym cyanoacetic acid,acetic acid, cyano,malonic mononitrile,acide cyanacetique,cyanessigsaeure,monocyanoacetic acid,malonic acid mononitrile,usaf kf-17,cyanoethanoic acid,kyselina kyanoctova
Solubility Information Miscible with water,ethyl alcohol,ether. Slightly miscible with benzene,acetic acid and chloroform.
InChI Key MLIREBYILWEBDM-UHFFFAOYSA-N
SMILES C(C#N)C(=O)O
IUPAC Name 2-cyanoacetic acid
Molecular Weight (g/mol) 85.062
PubChem CID 9740
ChEBI CHEBI:51889
Formula Weight 85.06
Percent Purity 99%
Applications
Cyanoacetic acid is used in pharmaceutical industry for producing vitamin B6 and caffeine. It is also used in making dyes, agricultural chemicals, and in the synthesis of phenylacetic acid esters. It acts as a precursor cyanoacrylates viz. ethyl cyanoacrylate.
Solubility
Miscible with water, ethyl alcohol, ether. Slightly miscible with benzene, acetic acid and chloroform.
Notes
Hygroscopic. Incompatible with strong bases, acids, strong oxidizing agents, alkaline, nitriles and reducing agents.