Acetone, 99+%, extra pure
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Overview
100.59 GBP valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Code: 29
Additional Details:
Additional Details: CAS Number: 67-64-1 Weight: 3.95000kg Transport: UN number : 1090 Chem class : 3 Pack group : II
Quantity 5 L
Packaging Plastic Drum
Disclaimers: RUO – Research Use Only
Product Code. 10745863
Quantity Price
1 £ 112.0 / 5 L
In stock, ready to ship: 8
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Chemical Identifiers
CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
Specification
Chemical Name or Material Acetone
Melting Point -95.0°C
CAS 67-64-1
CAS Min % 99.0
CAS Max % 100.0
Decomposition Information >4°C
Density 0.7900g/mL
Quantity 5 L
Flash Point −20°C
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.3585
Residue after Evaporation 0.005% max.
Assay Percent Range 99+%
Specific Gravity 0.79
Molecular Formula C3H6O
Linear Formula CH3COCH3
Vapor Pressure Vapor Pressure: 247mbar at 20°C
MDL Number MFCD00008765
Viscosity 0.32 mPa.s (20°C)
UN Number 1090; Colorless
Water 0.5% max. (K.F.)
Beilstein 01, 635
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Synonym acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
Boiling Point 56.0°C
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Formula Weight 58.08
Percent Purity ≥99%
Grade Extra Pure
Alternative Products
Documentation Safety and Handling
[Acetone]
[Danger]
[{code=FLAM. LIQ. 2, sequence=1, description=Flammable liquid Category 2}, {code=EYE DAM. 2, sequence=2, description=Serious eye damage/eye irritation Category 2}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=EUH066, sequence=91, description=Repeated exposure may cause skin dryness or cracking.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P303+P361+P353, sequence=52, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P280, sequence=53, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P337+P313, sequence=54, description=If eye irritation persists: Get medical advice/attention.}, {code=P304+P340, sequence=55, description=IF INHALED: Remove person to fresh air and keep comfortable for breathing.}, {code=P312, sequence=56, description=Call a POISON CENTER/doctor/if you feel unwell.}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225- Highly flammable liquid and vapour.
  • H319- Causes serious eye irritation.
  • H336- May cause drowsiness or dizziness.
  • EUH066- Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313- If eye irritation persists: Get medical advice/attention.
  • P304+P340- IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312- Call a POISON CENTER/doctor/if you feel unwell.