2-Hydroxycyclohexanone dimer, 90+%
CAS: 30282-14-5 | C6H10O2 | 114.144 g/mol
Overview
107.99 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Code: NEW
Additional Details:
Additional Details: CAS Number: 30282-14-5 Weight: 0.02500kg Transport: UN number : 1325 Chem class : 4.1 Pack group : III
Quantity 25 g
Disclaimers: RUO – Research Use Only
Product Code. 11302179
Quantity Price
1 £ 108.0 / 25g
Estimated Shipment
19-02-2026
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Chemical Identifiers
CAS 30282-14-5
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00151157
InChI Key ODZTXUXIYGJLMC-UHFFFAOYSA-N
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
PubChem CID 10785
ChEBI CHEBI:17878
IUPAC Name 2-hydroxycyclohexan-1-one
SMILES C1CCC(=O)C(C1)O
Specification
Chemical Name or Material 2-Hydroxycyclohexanone dimer
CAS 30282-14-5
Melting Point 100°C to 107°C
Boiling Point 83°C to 86°C (13 mmHg)
Flash Point 79°C (174°F)
Molecular Formula C6H10O2
MDL Number MFCD00151157
Quantity 25 g
UN Number UN1325
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Synonym 2-hydroxycyclohexanone,adipoin,2-hydroxy-1-cyclohexanone,cyclohexanone, 2-hydroxy,2-hydroxy-cyclohexanone,cyclohexanone, 2-hydroxy-, 2r,cyclohexanone, 2-hydroxy-, dimer,cyclohexanone, dimer,zlchem 1128,acmc-20n5uh
Solubility Information Partly miscible in water.
InChI Key ODZTXUXIYGJLMC-UHFFFAOYSA-N
SMILES C1CCC(=O)C(C1)O
IUPAC Name 2-hydroxycyclohexan-1-one
Molecular Weight (g/mol) 114.144
PubChem CID 10785
ChEBI CHEBI:17878
Formula Weight 228.28
Percent Purity ≥90%
Applications
It is a reactant involved in the synthesis of substrates for tin tetrachloride mediated fragmentation to produce ynones and ynoates, tandem oxidation coupling reactions with diamines, asymmetric allylic alkylation of enolates and formate reduction of allylic carbonates for polypropionate systems.
Solubility
Partly miscible in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges. Ensure adequate ventilation.