CAS RN 99986-02-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl (5-Ethyl-2-pyridinyl)acetate, TRC
CAS: 99986-02-4 Molecular Formula: C11 H15 N O2 Molecular Weight (g/mol): 193.24 Synonym: 2-Pyridineacetic acid, 5-ethyl-, ethyl ester,5-Ethyl-2-pyridineacetic acid ethyl ester,Ethyl (5-ethyl-2-pyridinyl)acetate IUPAC Name: ethyl 2-(5-ethylpyridin-2-yl)acetate SMILES: CCOC(=O)Cc1ccc(CC)cn1