CAS RN 99073-95-7
IUPAC Name:
4-(2-bromophenyl)pyrimidin-2-amine
Synonyms:
4-(2-bromophenyl)pyrimidin-2-amine
Molecular Weight (g/mol):
250.1
Molecular Formula:
C10H8BrN3
InChi Key:
ZCMWKKYYDONJDU-UHFFFAOYSA-N
SMILES:
NC1=NC=CC(=N1)C1=CC=CC=C1Br
1
–
1
of
1
results
1
2-Amino-4-(2-bromophenyl)pyrimidine, 95%, Thermo Scientific™
CAS: 99073-95-7 Molecular Formula: C10H8BrN3 Molecular Weight (g/mol): 250.10 MDL Number: MFCD07381905 InChI Key: ZCMWKKYYDONJDU-UHFFFAOYSA-N Synonym: 4-2-bromophenyl pyrimidin-2-amine,2-amino-4-2-bromophenyl pyrimidine,4-2-bromophenyl-2-pyrimidinylamine PubChem CID: 53426180 IUPAC Name: 4-(2-bromophenyl)pyrimidin-2-amine SMILES: NC1=NC=CC(=N1)C1=CC=CC=C1Br