CAS RN 98930-01-9
CAS RN 98930-01-9
IUPAC Name:
5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms:
5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol):
608.71
Molecular Formula:
C31H36N4O7S
InChi Key:
LKGHIEITYHYVED-UHFFFAOYNA-N
SMILES:
COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
1
of
1
results
1
Nalpha-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine, 95%, Thermo Scientific™
CAS: 98930-01-9 Molecular Formula: C31H36N4O7S Molecular Weight (g/mol): 608.71 MDL Number: MFCD00043098,MFCD00065616 InChI Key: LKGHIEITYHYVED-UHFFFAOYNA-N Synonym: fmoc-arg mtr-oh,fmoc-l-arg mtr,fmoc-arg mtr,fmoc-l-arg mtr-oh,ambotzfaa1700,fmoc-arginine mtr-oh,fmoc-l-arginine mtr,pubchem10000,fmoc-ng-mtr-l-arginine hemiisopropyl ether,fmoc-arg mtr-oh tlc PubChem CID: 3010428 SMILES: COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O